Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAAGGCAGATTTTTCCAGAGATA[A/G]CAAAGACACTTTGTCACTTTGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609391 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01124 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC01124 - long intergenic non-protein coding RNA 1124 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101926913 - uncharacterized LOC101926913 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SP5 - Sp5 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003845.2 | 167 | Intron | NP_001003845.1 | |||
XM_005246542.4 | 167 | UTR 5 | XP_005246599.1 | |||
XM_011511159.2 | 167 | UTR 5 | XP_011509461.1 |
Set Membership: |
HapMap |