Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605541 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VAV3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VAV3 - vav guanine nucleotide exchange factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079874.1 | 3571 | UTR 3 | NP_001073343.1 | |||
NM_006113.4 | 3571 | UTR 3 | NP_006104.4 | |||
XM_005270360.2 | 3571 | UTR 3 | XP_005270417.1 | |||
XM_005270361.1 | 3571 | Intron | XP_005270418.1 | |||
XM_017000053.1 | 3571 | UTR 3 | XP_016855542.1 | |||
XM_017000054.1 | 3571 | Intron | XP_016855543.1 | |||
XM_017000055.1 | 3571 | UTR 3 | XP_016855544.1 | |||
XM_017000056.1 | 3571 | UTR 3 | XP_016855545.1 |