Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTAGGGGCACAGTGAACTATAT[C/T]AGTGAATAGATGGATGCACAGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616650 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSG1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GSG1L - GSG1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109763.1 | 3286 | UTR 3 | NP_001103233.1 | |||
NM_001323900.1 | 3286 | UTR 3 | NP_001310829.1 | |||
NM_001323901.1 | 3286 | UTR 3 | NP_001310830.1 | |||
NM_144675.2 | 3286 | UTR 3 | NP_653276.1 | |||
XM_011545750.2 | 3286 | Intron | XP_011544052.1 | |||
XM_011545751.2 | 3286 | Intron | XP_011544053.1 |
KIAA0556 - KIAA0556 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |