Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAACCTCAGATGCCTACAGCAG[C/T]CAGACAGAGCATACAAAAGCATGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600760 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCNN1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SCNN1B - sodium channel epithelial 1 beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000336.2 | Intron | NP_000327.2 | ||||
XM_011545913.2 | Intron | XP_011544215.1 | ||||
XM_011545914.1 | Intron | XP_011544216.1 | ||||
XM_017023525.1 | Intron | XP_016879014.1 | ||||
XM_017023526.1 | Intron | XP_016879015.1 |
Set Membership: |
HapMap |