Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF780B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF780B - zinc finger protein 780B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005851.2 | Intron | NP_001005851.1 | ||||
XM_005258590.4 | Intron | XP_005258647.1 | ||||
XM_005258591.4 | Intron | XP_005258648.1 | ||||
XM_005258592.3 | Intron | XP_005258649.1 | ||||
XM_005258593.2 | Intron | XP_005258650.1 | ||||
XM_005258595.3 | Intron | XP_005258652.1 | ||||
XM_006723072.3 | Intron | XP_006723135.1 | ||||
XM_011526593.2 | Intron | XP_011524895.1 | ||||
XM_017026425.1 | Intron | XP_016881914.1 | ||||
XM_017026426.1 | Intron | XP_016881915.1 | ||||
XM_017026427.1 | Intron | XP_016881916.1 |