Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAACAAAATGGTGGACTTCTTA[C/T]TAATGGTCACAGATAAATCAACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179730 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RLN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RLN1 - relaxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006911.3 | Intron | NP_008842.1 | ||||
XM_017015007.1 | Intron | XP_016870496.1 | ||||
XM_017015008.1 | Intron | XP_016870497.1 | ||||
XM_017015009.1 | Intron | XP_016870498.1 | ||||
XM_017015010.1 | Intron | XP_016870499.1 |
Set Membership: |
HapMap |