Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 400033 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBL1Y PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TBL1Y - transducin (beta)-like 1, Y-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033284.1 | Intron | NP_150600.1 | ||||
NM_134258.1 | Intron | NP_599020.1 | ||||
NM_134259.1 | Intron | NP_599021.1 | ||||
XM_005262572.3 | Intron | XP_005262629.1 | ||||
XM_017030086.1 | Intron | XP_016885575.1 | ||||
XM_017030087.1 | Intron | XP_016885576.1 |