Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCAGGGTTCTTCTAAGGATTCAG[C/T]GAAATAACATTAGAAACTCACAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611305 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABLIM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABLIM3 - actin binding LIM protein family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301015.1 | Intron | NP_001287944.1 | ||||
NM_001301018.1 | Intron | NP_001287947.1 | ||||
NM_001301027.1 | Intron | NP_001287956.1 | ||||
NM_001301028.1 | Intron | NP_001287957.1 | ||||
NM_014945.3 | Intron | NP_055760.1 | ||||
XM_005268392.1 | Intron | XP_005268449.1 | ||||
XM_005268393.1 | Intron | XP_005268450.1 | ||||
XM_005268394.1 | Intron | XP_005268451.1 | ||||
XM_005268395.1 | Intron | XP_005268452.1 | ||||
XM_005268396.1 | Intron | XP_005268453.1 | ||||
XM_011537609.2 | Intron | XP_011535911.1 | ||||
XM_017009238.1 | Intron | XP_016864727.1 |