Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611596 MIM: 616925 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DHRS4 - dehydrogenase/reductase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282987.1 | Intron | NP_001269916.1 | ||||
NM_001282988.1 | Intron | NP_001269917.1 | ||||
NM_001282989.1 | Intron | NP_001269918.1 | ||||
NM_001282990.1 | Intron | NP_001269919.1 | ||||
NM_001282991.1 | Intron | NP_001269920.1 | ||||
NM_021004.3 | Intron | NP_066284.2 | ||||
XM_006720005.3 | Intron | XP_006720068.1 | ||||
XM_011536366.2 | Intron | XP_011534668.1 |
DHRS4-AS1 - DHRS4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |