Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTGTGGCCAAAGAGGAGATTCT[A/G]GCCAATCTCTAAAGTGAAGGAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602565 MIM: 608529 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL25 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCL25 - C-C motif chemokine ligand 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBN3 - fibrillin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321431.1 | Intron | NP_001308360.1 | ||||
NM_032447.4 | Intron | NP_115823.3 | ||||
XM_017027372.1 | Intron | XP_016882861.1 | ||||
XM_017027373.1 | Intron | XP_016882862.1 | ||||
XM_017027374.1 | Intron | XP_016882863.1 | ||||
XM_017027375.1 | Intron | XP_016882864.1 | ||||
XM_017027376.1 | Intron | XP_016882865.1 | ||||
XM_017027377.1 | Intron | XP_016882866.1 | ||||
XM_017027378.1 | Intron | XP_016882867.1 | ||||
XM_017027379.1 | Intron | XP_016882868.1 |
Set Membership: |
HapMap |