Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAGAGCACGTGTGGTTAGAGGT[C/G]GGGGGGTAATCTTTTAACCTCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609405 | ||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP8 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ARHGAP8 - Rho GTPase activating protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR5-ARHGAP8 - PRR5-ARHGAP8 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |