Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611163 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TOX2 - TOX high mobility group box family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098796.1 | Intron | NP_001092266.1 | ||||
NM_001098797.1 | Intron | NP_001092267.1 | ||||
NM_001098798.1 | Intron | NP_001092268.1 | ||||
NM_032883.2 | Intron | NP_116272.1 | ||||
XM_006723884.1 | Intron | XP_006723947.1 | ||||
XM_017028109.1 | Intron | XP_016883598.1 |