Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGACTTTGCACTTGGATGTGAAA[C/T]CTGCTACTCAGTCCTTCAGGCTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611163 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOX2 - TOX high mobility group box family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098796.1 | Intron | NP_001092266.1 | ||||
NM_001098797.1 | Intron | NP_001092267.1 | ||||
NM_001098798.1 | Intron | NP_001092268.1 | ||||
NM_032883.2 | Intron | NP_116272.1 | ||||
XM_006723884.1 | Intron | XP_006723947.1 | ||||
XM_017028109.1 | Intron | XP_016883598.1 |