Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608489 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GATS - GATS, stromal antigen 3 opposite strand | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPC2 - glypican 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAG3 - stromal antigen 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282716.1 | Intron | NP_001269645.1 | ||||
NM_001282717.1 | Intron | NP_001269646.1 | ||||
NM_001282718.1 | Intron | NP_001269647.1 | ||||
NM_012447.3 | Intron | NP_036579.2 | ||||
XM_011515742.1 | Intron | XP_011514044.1 | ||||
XM_017011683.1 | Intron | XP_016867172.1 | ||||
XM_017011684.1 | Intron | XP_016867173.1 | ||||
XM_017011685.1 | Intron | XP_016867174.1 | ||||
XM_017011686.1 | Intron | XP_016867175.1 | ||||
XM_017011687.1 | Intron | XP_016867176.1 |
Set Membership: |
HapMap |