Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTTTATTACCAAATCCTTTCAT[C/T]AGGTAGTACCAAGTAAACAGCATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606902 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107984745 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC107984745 - uncharacterized mitochondrial protein AtMg00860-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYGO1 - pygopus family PHD finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015617.2 | 7490 | UTR 3 | NP_056432.1 | |||
XM_005254300.4 | 7490 | UTR 3 | XP_005254357.1 | |||
XM_011521446.2 | 7490 | UTR 3 | XP_011519748.1 |
Set Membership: |
HapMap |