Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATAATAAAAAATCCAAGTAAAA[C/T]GAGATCTTCAAGTACACTACTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PYGO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PYGO1 - pygopus family PHD finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015617.2 | 3744 | UTR 3 | NP_056432.1 | |||
XM_005254300.4 | 3744 | UTR 3 | XP_005254357.1 | |||
XM_011521446.2 | 3744 | UTR 3 | XP_011519748.1 |