Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACATAATTATGTATCCTGTGTTT[A/G]TTTATTTGGCTCTCCATCTTGAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616578 | ||||||||||||||||||||
Literature Links: |
LOC102723809 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC102723809 - uncharacterized LOC102723809 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB1 - zinc finger and BTB domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123329.1 | Intron | NP_001116801.1 | ||||
NM_014950.2 | Intron | NP_055765.2 | ||||
XM_005267410.1 | Intron | XP_005267467.1 | ||||
XM_011536566.2 | Intron | XP_011534868.1 | ||||
XM_011536568.2 | Intron | XP_011534870.1 | ||||
XM_017021094.1 | Intron | XP_016876583.1 | ||||
XM_017021095.1 | Intron | XP_016876584.1 |