Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605304 MIM: 607439 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NGB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NGB - neuroglobin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POMT2 - protein O-mannosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013382.5 | 3346 | UTR 3 | NP_037514.2 | |||
XM_011536675.2 | 3346 | UTR 3 | XP_011534977.1 | |||
XM_011536676.2 | 3346 | UTR 3 | XP_011534978.1 | |||
XM_011536677.2 | 3346 | UTR 3 | XP_011534979.1 | |||
XM_011536679.2 | 3346 | UTR 3 | XP_011534981.1 | |||
XM_017021244.1 | 3346 | UTR 3 | XP_016876733.1 | |||
XM_017021245.1 | 3346 | UTR 3 | XP_016876734.1 |
Set Membership: |
HapMap |