Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTAGCGAATGCCACAGCCTGGG[C/T]GGCTCAGACGACAGCCAGTCACTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610285 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOK7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DOK7 - docking protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164673.1 | Intron | NP_001158145.1 | ||||
NM_001256896.1 | Intron | NP_001243825.1 | ||||
NM_001301071.1 | Intron | NP_001288000.1 | ||||
NM_173660.4 | Intron | NP_775931.3 | ||||
XM_011513435.2 | Intron | XP_011511737.1 | ||||
XM_011513437.2 | Intron | XP_011511739.1 | ||||
XM_017008038.1 | Intron | XP_016863527.1 |