Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTGTGAATGGAGACTGGTCCCAG[A/G]AAGTGGCAGCTCTCGGGACCGCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614620 MIM: 611002 | ||||||||||||||||||||
Literature Links: |
IFT140 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFT140 - intraflagellar transport 140 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014714.3 | Intron | NP_055529.2 |
LOC105371047 - uncharacterized LOC105371047 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM204 - transmembrane protein 204 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256541.1 | Intron | NP_001243470.1 | ||||
NM_024600.5 | Intron | NP_078876.2 |