Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCAAACTGAGCACAGGCTGAGC[C/T]TGATCTTCTTACCCGTTCATTCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107987125 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107987125 - uncharacterized LOC107987125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTLL11 - tubulin tyrosine ligase like 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139442.1 | 4997 | Intron | NP_001132914.1 | |||
NM_194252.2 | 4997 | Intron | NP_919228.2 | |||
XM_005251728.2 | 4997 | UTR 3 | XP_005251785.1 | |||
XM_005251729.4 | 4997 | UTR 3 | XP_005251786.1 | |||
XM_006716966.3 | 4997 | Intron | XP_006717029.1 | |||
XM_017014309.1 | 4997 | Intron | XP_016869798.1 |
Set Membership: |
HapMap |