Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGAGACTCCAGGTTTTTGGGGGG[G/T]TTTTTGCTTTTTTTAATAGAGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608465 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SETX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SETX - senataxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015046.5 | 3534 | UTR 3 | NP_055861.3 | |||
XM_005272171.1 | 3534 | UTR 3 | XP_005272228.1 | |||
XM_005272172.2 | 3534 | Intron | XP_005272229.1 | |||
XM_005272173.2 | 3534 | Intron | XP_005272230.1 | |||
XM_011518404.2 | 3534 | Intron | XP_011516706.1 | |||
XM_011518405.2 | 3534 | Intron | XP_011516707.1 | |||
XM_011518406.2 | 3534 | Intron | XP_011516708.1 | |||
XM_011518407.1 | 3534 | Intron | XP_011516709.1 | |||
XM_011518408.2 | 3534 | Intron | XP_011516710.1 | |||
XM_017014495.1 | 3534 | UTR 3 | XP_016869984.1 | |||
XM_017014496.1 | 3534 | UTR 3 | XP_016869985.1 | |||
XM_017014497.1 | 3534 | UTR 3 | XP_016869986.1 |
Set Membership: |
HapMap |