Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611104 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGD4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FGD4 - FYVE, RhoGEF and PH domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304480.1 | Intron | NP_001291409.1 | ||||
NM_001304481.1 | Intron | NP_001291410.1 | ||||
NM_001304483.1 | Intron | NP_001291412.1 | ||||
NM_001304484.1 | Intron | NP_001291413.1 | ||||
NM_139241.3 | Intron | NP_640334.2 | ||||
XM_005253304.4 | Intron | XP_005253361.1 | ||||
XM_005253307.3 | Intron | XP_005253364.1 | ||||
XM_005253308.4 | Intron | XP_005253365.1 | ||||
XM_005253309.1 | Intron | XP_005253366.1 | ||||
XM_005253310.3 | Intron | XP_005253367.1 | ||||
XM_011520554.1 | Intron | XP_011518856.1 | ||||
XM_011520555.1 | Intron | XP_011518857.1 | ||||
XM_011520556.1 | Intron | XP_011518858.1 | ||||
XM_011520557.1 | Intron | XP_011518859.1 | ||||
XM_011520558.2 | Intron | XP_011518860.1 | ||||
XM_011520559.2 | Intron | XP_011518861.1 | ||||
XM_017018803.1 | Intron | XP_016874292.1 | ||||
XM_017018804.1 | Intron | XP_016874293.1 | ||||
XM_017018805.1 | Intron | XP_016874294.1 |
Set Membership: |
HapMap |