Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTGGACATATATAGACCTGAT[G/T]ACCTTGGGCAAGTTATTCGACATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609174 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NSL1 - NSL1, MIS12 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042549.1 | Intron | NP_001036014.1 | ||||
NM_001297736.1 | Intron | NP_001284665.1 | ||||
NM_001297737.1 | Intron | NP_001284666.1 | ||||
NM_001297739.1 | Intron | NP_001284668.1 | ||||
NM_015471.3 | Intron | NP_056286.3 | ||||
XM_017000968.1 | Intron | XP_016856457.1 | ||||
XM_017000969.1 | Intron | XP_016856458.1 |
TATDN3 - TatD DNase domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042552.2 | Intron | NP_001036017.1 | ||||
NM_001042553.2 | Intron | NP_001036018.1 | ||||
NM_001146169.1 | Intron | NP_001139641.1 | ||||
NM_001146170.1 | Intron | NP_001139642.1 | ||||
NM_001146171.1 | Intron | NP_001139643.1 | ||||
XM_011509202.2 | Intron | XP_011507504.1 | ||||
XM_017000328.1 | Intron | XP_016855817.1 | ||||
XM_017000329.1 | Intron | XP_016855818.1 | ||||
XM_017000330.1 | Intron | XP_016855819.1 | ||||
XM_017000331.1 | Intron | XP_016855820.1 |
Set Membership: |
HapMap |