Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605347 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PADI4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PADI4 - peptidyl arginine deiminase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012387.2 | Intron | NP_036519.2 | ||||
XM_011541150.1 | Intron | XP_011539452.1 | ||||
XM_011541151.1 | Intron | XP_011539453.1 | ||||
XM_011541152.1 | Intron | XP_011539454.1 | ||||
XM_011541153.1 | Intron | XP_011539455.1 | ||||
XM_011541154.2 | Intron | XP_011539456.1 | ||||
XM_011541155.1 | Intron | XP_011539457.1 | ||||
XM_011541156.1 | Intron | XP_011539458.1 | ||||
XM_011541157.1 | Intron | XP_011539459.1 |