Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGCATGGACCAAGGCCCCTCCC[A/G]TAAATTACATTGTTAGCATAATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601953 MIM: 139150 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNH - cyclin H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199189.1 | Intron | NP_001186118.1 | ||||
NM_001239.3 | Intron | NP_001230.1 | ||||
XM_005248627.3 | Intron | XP_005248684.1 | ||||
XM_005248629.4 | Intron | XP_005248686.1 | ||||
XM_011543706.2 | Intron | XP_011542008.1 | ||||
XM_017010014.1 | Intron | XP_016865503.1 |
LOC644285 - uncharacterized LOC644285 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASA1 - RAS p21 protein activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002890.2 | Intron | NP_002881.1 | ||||
NM_022650.2 | Intron | NP_072179.1 | ||||
XM_011543525.1 | Intron | XP_011541827.1 | ||||
XM_011543527.2 | Intron | XP_011541829.1 |
Set Membership: |
HapMap |