Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGCTGCTTCCTCTACCCCTGTA[G/T]TTCGCTCTGCTGAGTCTGACTTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613541 MIM: 602198 MIM: 605501 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf65 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C12orf65 - chromosome 12 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143905.2 | Intron | NP_001137377.1 | ||||
NM_001194995.1 | Intron | NP_001181924.1 | ||||
NM_152269.4 | Intron | NP_689482.1 | ||||
XM_005253630.4 | Intron | XP_005253687.1 | ||||
XM_011538980.2 | Intron | XP_011537282.1 |
CDK2AP1 - cyclin dependent kinase 2 associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPHOSPH9 - M-phase phosphoprotein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |