Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSHR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FSHR - follicle stimulating hormone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000145.3 | 4542 | Intron | NP_000136.2 | |||
NM_181446.2 | 4542 | Intron | NP_852111.2 | |||
XM_011532733.2 | 4542 | UTR 3 | XP_011531035.1 | |||
XM_011532734.2 | 4542 | UTR 3 | XP_011531036.1 | |||
XM_011532735.2 | 4542 | UTR 3 | XP_011531037.1 | |||
XM_011532736.2 | 4542 | UTR 3 | XP_011531038.1 | |||
XM_011532740.1 | 4542 | Intron | XP_011531042.1 |