Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616297 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCTP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MCTP2 - multiple C2 and transmembrane domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159643.1 | Intron | NP_001153115.1 | ||||
NM_001159644.1 | Intron | NP_001153116.1 | ||||
NM_018349.3 | Intron | NP_060819.3 | ||||
XM_005254955.3 | Intron | XP_005255012.1 | ||||
XM_005254960.2 | Intron | XP_005255017.1 | ||||
XM_006720603.2 | Intron | XP_006720666.1 | ||||
XM_011521770.1 | Intron | XP_011520072.1 | ||||
XM_011521771.2 | Intron | XP_011520073.1 | ||||
XM_011521772.2 | Intron | XP_011520074.1 | ||||
XM_011521773.2 | Intron | XP_011520075.1 | ||||
XM_011521774.2 | Intron | XP_011520076.1 | ||||
XM_011521775.2 | Intron | XP_011520077.1 | ||||
XM_017022403.1 | Intron | XP_016877892.1 | ||||
XM_017022404.1 | Intron | XP_016877893.1 | ||||
XM_017022405.1 | Intron | XP_016877894.1 |