Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153245 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LEF1 - lymphoid enhancer binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130713.2 | Intron | NP_001124185.1 | ||||
NM_001130714.2 | Intron | NP_001124186.1 | ||||
NM_001166119.1 | Intron | NP_001159591.1 | ||||
NM_016269.4 | Intron | NP_057353.1 | ||||
XM_005263046.3 | Intron | XP_005263103.1 | ||||
XM_005263047.1 | Intron | XP_005263104.1 | ||||
XM_005263048.1 | Intron | XP_005263105.1 | ||||
XM_006714233.1 | Intron | XP_006714296.1 |
Set Membership: |
HapMap |