Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 616476 | ||||||||||||||||||||||||||
Literature Links: |
AGBL4 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AGBL4 - ATP/GTP binding protein like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323573.1 | 2894 | UTR 3 | NP_001310502.1 | |||
NM_001323574.1 | 2894 | UTR 3 | NP_001310503.1 | |||
NM_001323575.1 | 2894 | UTR 3 | NP_001310504.1 | |||
NM_032785.3 | 2894 | UTR 3 | NP_116174.3 | |||
XM_011542308.2 | 2894 | UTR 3 | XP_011540610.1 | |||
XM_011542310.2 | 2894 | Intron | XP_011540612.1 | |||
XM_017002595.1 | 2894 | Intron | XP_016858084.1 | |||
XM_017002596.1 | 2894 | Intron | XP_016858085.1 | |||
XM_017002597.1 | 2894 | UTR 3 | XP_016858086.1 | |||
XM_017002598.1 | 2894 | Intron | XP_016858087.1 |
Set Membership: |
HapMap |