Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCTTCGATGCGGACCTTCTGG[T/G]TTTTGATCTCCTCGATGTCCTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 121011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJB2 - gap junction protein beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004004.5 | 574 | Missense Mutation | AAC,ACC | N,T 123 | NP_003995.2 | |
XM_011535049.2 | 574 | Missense Mutation | AAC,ACC | N,T 123 | XP_011533351.1 |