Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608777 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POSTN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POSTN - periostin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135934.1 | 2918 | Intron | NP_001129406.1 | |||
NM_001135935.1 | 2918 | Intron | NP_001129407.1 | |||
NM_001135936.1 | 2918 | Intron | NP_001129408.1 | |||
NM_001286665.1 | 2918 | Intron | NP_001273594.1 | |||
NM_001286666.1 | 2918 | Intron | NP_001273595.1 | |||
NM_001286667.1 | 2918 | Intron | NP_001273596.1 | |||
NM_006475.2 | 2918 | Intron | NP_006466.2 | |||
XM_005266231.2 | 2918 | Intron | XP_005266288.1 | |||
XM_005266232.2 | 2918 | Intron | XP_005266289.1 | |||
XM_017020355.1 | 2918 | Intron | XP_016875844.1 | |||
XM_017020356.1 | 2918 | UTR 3 | XP_016875845.1 |
Set Membership: |
HapMap |