Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTCCTCTAATCAAAAATGAAAA[C/G]AAGACCAAAAACGACTAACTTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607796 MIM: 607867 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHF11 - PHD finger protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCBTB1 - RCC1 and BTB domain containing protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018191.3 | 2068 | UTR 3 | NP_060661.3 | |||
XM_005266441.2 | 2068 | UTR 3 | XP_005266498.1 | |||
XM_011535133.1 | 2068 | UTR 3 | XP_011533435.1 | |||
XM_011535134.1 | 2068 | UTR 3 | XP_011533436.1 | |||
XM_011535135.2 | 2068 | UTR 3 | XP_011533437.1 | |||
XM_011535136.2 | 2068 | Intron | XP_011533438.1 |
SETDB2-PHF11 - SETDB2-PHF11 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |