Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614289 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEL1L2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SEL1L2 - SEL1L2 ERAD E3 ligase adaptor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271539.1 | Intron | NP_001258468.1 | ||||
XM_006723646.2 | Intron | XP_006723709.1 | ||||
XM_006723647.2 | Intron | XP_006723710.1 | ||||
XM_006723648.2 | Intron | XP_006723711.1 | ||||
XM_006723650.2 | Intron | XP_006723713.1 | ||||
XM_006723651.2 | Intron | XP_006723714.1 | ||||
XM_006723652.2 | Intron | XP_006723715.1 | ||||
XM_006723654.3 | Intron | XP_006723717.1 | ||||
XM_006723655.2 | Intron | XP_006723718.1 | ||||
XM_006723656.2 | Intron | XP_006723719.1 | ||||
XM_011529374.1 | Intron | XP_011527676.1 | ||||
XM_011529375.1 | Intron | XP_011527677.1 | ||||
XM_011529376.1 | Intron | XP_011527678.1 | ||||
XM_011529377.1 | Intron | XP_011527679.1 | ||||
XM_011529378.1 | Intron | XP_011527680.1 | ||||
XM_011529379.1 | Intron | XP_011527681.1 | ||||
XM_017028084.1 | Intron | XP_016883573.1 |