Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611120 MIM: 608270 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPTLC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPTLC3 - serine palmitoyltransferase long chain base subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018327.2 | Intron | NP_060797.2 | ||||
XM_011529279.1 | Intron | XP_011527581.1 | ||||
XM_011529280.2 | Intron | XP_011527582.1 | ||||
XM_017027922.1 | Intron | XP_016883411.1 |
TASP1 - taspase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323602.1 | Intron | NP_001310531.1 | ||||
NM_001323603.1 | Intron | NP_001310532.1 | ||||
NM_001323604.1 | Intron | NP_001310533.1 | ||||
NM_017714.2 | Intron | NP_060184.2 | ||||
XM_011529292.1 | Intron | XP_011527594.1 | ||||
XM_011529294.1 | Intron | XP_011527596.1 | ||||
XM_017027927.1 | Intron | XP_016883416.1 | ||||
XM_017027928.1 | Intron | XP_016883417.1 | ||||
XM_017027929.1 | Intron | XP_016883418.1 | ||||
XM_017027930.1 | Intron | XP_016883419.1 | ||||
XM_017027931.1 | Intron | XP_016883420.1 |
Set Membership: |
HapMap |