Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAGAGAATAATAGTGTGATAT[A/G]TGAAAAGCCAAAAGAAAAAAAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610236 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LNPK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LNPK - lunapark, ER junction formation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305008.1 | 4441 | UTR 3 | NP_001291937.1 | |||
NM_001305009.1 | 4441 | UTR 3 | NP_001291938.1 | |||
NM_001305010.1 | 4441 | UTR 3 | NP_001291939.1 | |||
NM_001305011.1 | 4441 | UTR 3 | NP_001291940.1 | |||
NM_030650.2 | 4441 | UTR 3 | NP_085153.1 | |||
XM_005246878.3 | 4441 | UTR 3 | XP_005246935.1 | |||
XM_005246879.4 | 4441 | UTR 3 | XP_005246936.1 | |||
XM_005246880.3 | 4441 | UTR 3 | XP_005246937.1 | |||
XM_006712783.2 | 4441 | UTR 3 | XP_006712846.1 | |||
XM_011511945.2 | 4441 | UTR 3 | XP_011510247.1 | |||
XM_017005055.1 | 4441 | UTR 3 | XP_016860544.1 |