Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTTTATTTCACAAAACTTCTTC[A/G]TTTCCAAATCTTATCTGTGCTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609479 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLK7-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLK7-AS1 - MLK7 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZAK - sterile alpha motif and leucine zipper containing kinase AZK | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016653.2 | Intron | NP_057737.2 | ||||
NM_133646.2 | Intron | NP_598407.1 | ||||
XM_005246640.2 | Intron | XP_005246697.1 | ||||
XM_017004323.1 | Intron | XP_016859812.1 | ||||
XM_017004324.1 | Intron | XP_016859813.1 |