Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATGACACTGGGTTAAGGAGCCA[C/T]CATCTACACTTAAAACCACTGCCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 309060 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LAMP2 - lysosomal associated membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122606.1 | 3808 | Intron | NP_001116078.1 | |||
NM_002294.2 | 3808 | UTR 3 | NP_002285.1 | |||
NM_013995.2 | 3808 | Intron | NP_054701.1 |
LOC105373326 - uncharacterized LOC105373326 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |