Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGCTAGTTTTCCTTATAACCAC[C/T]TCCTGGGCTGCTAGGCCACCTCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613992 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R2D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP2R2D - protein phosphatase 2 regulatory subunit Bdelta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291310.1 | Intron | NP_001278239.1 | ||||
NM_018461.4 | Intron | NP_060931.2 | ||||
XM_006717914.3 | Intron | XP_006717977.1 | ||||
XM_006717915.2 | Intron | XP_006717978.1 | ||||
XM_017016406.1 | Intron | XP_016871895.1 | ||||
XM_017016407.1 | Intron | XP_016871896.1 | ||||
XM_017016408.1 | Intron | XP_016871897.1 |
Set Membership: |
HapMap |