Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTGAGAAAATTCTCCTAAAACC[A/G]TCTCAAAAATAATTTAAAAAGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCML4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SCML4 - sex comb on midleg-like 4 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286408.1 | 5421 | Intron | NP_001273337.1 | |||
NM_001286409.1 | 5421 | Intron | NP_001273338.1 | |||
NM_198081.4 | 5421 | Intron | NP_932347.2 | |||
XM_011535703.2 | 5421 | UTR 3 | XP_011534005.1 | |||
XM_011535704.2 | 5421 | UTR 3 | XP_011534006.1 | |||
XM_011535706.1 | 5421 | UTR 3 | XP_011534008.1 | |||
XM_011535707.2 | 5421 | Intron | XP_011534009.1 | |||
XM_011535709.2 | 5421 | UTR 3 | XP_011534011.1 | |||
XM_011535710.2 | 5421 | UTR 3 | XP_011534012.1 | |||
XM_017010677.1 | 5421 | UTR 3 | XP_016866166.1 | |||
XM_017010678.1 | 5421 | UTR 3 | XP_016866167.1 | |||
XM_017010679.1 | 5421 | UTR 3 | XP_016866168.1 | |||
XM_017010680.1 | 5421 | UTR 3 | XP_016866169.1 | |||
XM_017010681.1 | 5421 | UTR 3 | XP_016866170.1 | |||
XM_017010682.1 | 5421 | UTR 3 | XP_016866171.1 | |||
XM_017010683.1 | 5421 | UTR 3 | XP_016866172.1 | |||
XM_017010684.1 | 5421 | Intron | XP_016866173.1 | |||
XM_017010685.1 | 5421 | UTR 3 | XP_016866174.1 | |||
XM_017010686.1 | 5421 | UTR 3 | XP_016866175.1 | |||
XM_017010687.1 | 5421 | UTR 3 | XP_016866176.1 | |||
XM_017010688.1 | 5421 | Intron | XP_016866177.1 |
Set Membership: |
HapMap |