Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGATGTTGAATATTGTCGGTTGA[C/T]GTGGTCTTGTTGCAGATTACATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf99 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C6orf99 - chromosome 6 open reading frame 99 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195032.1 | Intron | NP_001181961.1 | ||||
NM_001302839.1 | Intron | NP_001289768.1 | ||||
NM_001302840.1 | Intron | NP_001289769.1 | ||||
NM_001302841.1 | Intron | NP_001289770.1 | ||||
XM_005266788.4 | Intron | XP_005266845.1 | ||||
XM_011535374.2 | Intron | XP_011533676.1 | ||||
XM_017010142.1 | Intron | XP_016865631.1 | ||||
XM_017010143.1 | Intron | XP_016865632.1 | ||||
XM_017010144.1 | Intron | XP_016865633.1 | ||||
XM_017010145.1 | Intron | XP_016865634.1 | ||||
XM_017010146.1 | Intron | XP_016865635.1 | ||||
XM_017010147.1 | Intron | XP_016865636.1 |