Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAATCAGCATTACAATGTTAATT[C/T]TTTTTCTCTTAATATTTGTTTCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602190 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EPHA7 - EPH receptor A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288629.1 | 5816 | UTR 3 | NP_001275558.1 | |||
NM_001288630.1 | 5816 | Intron | NP_001275559.1 | |||
NM_004440.3 | 5816 | UTR 3 | NP_004431.1 | |||
XM_005248669.2 | 5816 | UTR 3 | XP_005248726.1 | |||
XM_005248671.2 | 5816 | Intron | XP_005248728.1 | |||
XM_017010365.1 | 5816 | UTR 3 | XP_016865854.1 | |||
XM_017010366.1 | 5816 | Intron | XP_016865855.1 |
Set Membership: |
HapMap |