Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGACGGGCTTAAATGCGTCTCC[A/G]CTGTGTTTCTTATGCCTCGGGCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616527 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP43 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CFAP43 - cilia and flagella associated protein 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFR1 - SWI5 dependent homologous recombination repair protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002759.1 | 201 | Intron | NP_001002759.1 | |||
NM_145247.4 | 201 | Intron | NP_660290.3 | |||
XM_005269521.3 | 201 | UTR 5 | XP_005269578.1 | |||
XM_017015672.1 | 201 | Intron | XP_016871161.1 |
Set Membership: |
HapMap |