Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614358 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSM2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSM2A - acyl-CoA synthetase medium-chain family member 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308169.1 | Intron | NP_001295098.1 | ||||
NM_001308172.1 | Intron | NP_001295101.1 | ||||
NM_001308954.1 | Intron | NP_001295883.1 | ||||
XM_017022923.1 | Intron | XP_016878412.1 | ||||
XM_017022924.1 | Intron | XP_016878413.1 | ||||
XM_017022925.1 | Intron | XP_016878414.1 | ||||
XM_017022926.1 | Intron | XP_016878415.1 |