Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604473 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRM1 - glutamate metabotropic receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278064.1 | Intron | NP_001264993.1 | ||||
NM_001278065.1 | Intron | NP_001264994.1 | ||||
NM_001278066.1 | Intron | NP_001264995.1 | ||||
NM_001278067.1 | Intron | NP_001264996.1 | ||||
XM_011535782.1 | Intron | XP_011534084.1 | ||||
XM_017010783.1 | Intron | XP_016866272.1 | ||||
XM_017010784.1 | Intron | XP_016866273.1 | ||||
XM_017010785.1 | Intron | XP_016866274.1 | ||||
XM_017010786.1 | Intron | XP_016866275.1 | ||||
XM_017010787.1 | Intron | XP_016866276.1 | ||||
XM_017010788.1 | Intron | XP_016866277.1 |
Set Membership: |
HapMap |