Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTCTGCCCTCCCCTCCCCTCC[G/T]CTACAGCCTCCAGAGTTAGGTAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105378159 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105378159 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017011599.1 | Intron | XP_016867088.1 |
SAMD3 - sterile alpha motif domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM200A - transmembrane protein 200A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258276.1 | Intron | NP_001245205.1 | ||||
NM_001258277.1 | Intron | NP_001245206.1 | ||||
NM_001258278.1 | Intron | NP_001245207.1 | ||||
NM_052913.2 | Intron | NP_443145.1 | ||||
XM_005266816.2 | Intron | XP_005266873.1 | ||||
XM_011535419.2 | Intron | XP_011533721.1 | ||||
XM_017010230.1 | Intron | XP_016865719.1 |