Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604334 MIM: 616463 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USP6 - ubiquitin specific peptidase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304284.1 | Intron | NP_001291213.1 | ||||
NM_004505.3 | Intron | NP_004496.2 | ||||
XM_011524050.1 | Intron | XP_011522352.1 | ||||
XM_011524051.2 | Intron | XP_011522353.1 | ||||
XM_011524052.2 | Intron | XP_011522354.1 | ||||
XM_011524053.2 | Intron | XP_011522355.1 | ||||
XM_011524054.2 | Intron | XP_011522356.1 | ||||
XM_011524055.2 | Intron | XP_011522357.1 | ||||
XM_011524056.2 | Intron | XP_011522358.1 | ||||
XM_011524057.2 | Intron | XP_011522359.1 | ||||
XM_011524058.2 | Intron | XP_011522360.1 | ||||
XM_011524059.2 | Intron | XP_011522361.1 | ||||
XM_017025290.1 | Intron | XP_016880779.1 |
ZNF232 - zinc finger protein 232 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |