Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605348 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSRP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RSRP1 - arginine and serine rich protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM50A - transmembrane protein 50A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014313.3 | 345 | Missense Mutation | GCC,GTC | A,V 58 | NP_055128.1 | |
XM_005245817.1 | 345 | Missense Mutation | GCC,GTC | A,V 58 | XP_005245874.1 | |
XM_011541159.1 | 345 | Missense Mutation | GCC,GTC | A,V 58 | XP_011539461.1 |