Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600408 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDR1 - discoidin domain receptor tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202521.1 | 6 | Intron | NP_001189450.1 | |||
NM_001202522.1 | 6 | Intron | NP_001189451.1 | |||
NM_001202523.1 | 6 | Intron | NP_001189452.1 | |||
NM_001297652.1 | 6 | UTR 5 | NP_001284581.1 | |||
NM_001297653.1 | 6 | Intron | NP_001284582.1 | |||
NM_001297654.1 | 6 | Intron | NP_001284583.1 | |||
NM_001954.4 | 6 | Intron | NP_001945.3 | |||
NM_013993.2 | 6 | Intron | NP_054699.2 | |||
NM_013994.2 | 6 | Intron | NP_054700.2 | |||
XM_006715185.2 | 6 | Intron | XP_006715248.1 | |||
XM_011514882.2 | 6 | Intron | XP_011513184.1 | |||
XM_011514883.1 | 6 | Intron | XP_011513185.1 | |||
XM_011514884.1 | 6 | Intron | XP_011513186.1 | |||
XM_011514885.1 | 6 | Intron | XP_011513187.1 | |||
XM_011514886.1 | 6 | Intron | XP_011513188.1 | |||
XM_011514887.1 | 6 | Intron | XP_011513189.1 | |||
XM_011514888.2 | 6 | Intron | XP_011513190.1 | |||
XM_017011268.1 | 6 | Intron | XP_016866757.1 | |||
XM_017011269.1 | 6 | Intron | XP_016866758.1 |
MIR4640 - microRNA 4640 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |